Skip to Content
Merck
All Photos(1)

Key Documents

80828

Supelco

Dimethylpropiothetin hydrochloride

PESTANAL®, analytical standard

Synonym(s):

DMSP, Dimethyl-3-propiothetin chloride, Dimethylsulfoniopropionate hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€55.70

€55.70


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€55.70

About This Item

Empirical Formula (Hill Notation):
C5H10O2S· HCl
CAS Number:
Molecular Weight:
170.66
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€55.70


Estimated to ship on28 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥96.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤5.0% water

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

[S+](CCC(=O)[O-])(C)C.Cl

InChI

1S/C5H10O2S.ClH/c1-8(2)4-3-5(6)7;/h3-4H2,1-2H3;1H

InChI key

RRUMKKGRKSSZKY-UHFFFAOYSA-N

General description

Dimethylpropiothetin is a natural product found in the organisms of saline environment such as algae and aquatic plants.[1]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Demethylation of dimethylsulfoniopropionate and production of thiols in anoxic marine sediments
Kiene PR and Taylor FB
Applied and Environmental Microbiology, 54(9), 2208-2212 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service