Skip to Content
Merck
All Photos(2)

Key Documents

D96006

Sigma-Aldrich

Diethyl glutarate

≥99%

Synonym(s):

Diethyl pentanedioate, Ethyl glutarate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€135.00

€135.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€135.00

About This Item

Linear Formula:
C2H5O2C(CH2)3CO2C2H5
CAS Number:
Molecular Weight:
188.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€135.00


Check Cart for Availability

Request a Bulk Order

Assay

≥99%

form

liquid

refractive index

n20/D 1.423 (lit.)

bp

237 °C (lit.)

density

1.022 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CCCC(=O)OCC

InChI

1S/C9H16O4/c1-3-12-8(10)6-5-7-9(11)13-4-2/h3-7H2,1-2H3

InChI key

OUWSNHWQZPEFEX-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.0 °F - closed cup

Flash Point(C)

95.56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J P Turner et al.
British journal of pharmacology, 104(2), 445-451 (1991-10-01)
1. The effects of L-glutamate diethyl ester (GDEE) HCl, glutarate diethyl ester (GlrDEE) and glutarate dimethyl ester (GlrDME) on depolarizing responses to alpha-amino-3-hydroxy-5- methyl-4-isoxazolepropionate (AMPA), kainate (Kain), N-methyl-D-aspartate (NMDA) and quisqualate (Quis), and spontaneous paroxysmal discharges (SPDs) were examined. Experiments
Ana-Marija Jagatić Korenika et al.
Molecules (Basel, Switzerland), 25(18) (2020-09-26)
Commercial sparkling wine production represents a relatively low but important part of the Croatian wine production, especially in the Zagreb county. This study presents the results of volatile aroma compounds profile and organic acid composition of commercial sparkling wine samples

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service