Skip to Content
Merck
All Photos(2)

Key Documents

804819

Sigma-Aldrich

Ammonium nitrate

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4NO3
CAS Number:
Molecular Weight:
80.04
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
powder or crystals
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

product line

Redi-Dri

Assay

≥98%

form

powder or crystals

quality

free-flowing

impurities

≤0.005% insolubles

ign. residue

≤0.01%

pH

4.5-6.0 (25 °C, 5%)

bp

210 °C (lit.)

mp

169 °C (lit.)

anion traces

chloride (Cl-): ≤5 ppm
nitrite (NO2-): passes test (lim. ∼5 ppm)
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.002%

cation traces

Fe: ≤2 ppm
heavy metals (as Pb): ≤5 ppm

SMILES string

N.O[N+]([O-])=O

InChI

1S/HNO3.H3N/c2-1(3)4;/h(H,2,3,4);1H3

InChI key

PRORZGWHZXZQMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1C - Ammonium nitrate and ammonium nitrate containing preparations

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah J Benson et al.
Science & justice : journal of the Forensic Science Society, 49(2), 73-80 (2009-07-18)
An evaluation was undertaken to determine if isotope ratio mass spectrometry (IRMS) could assist in the investigation of complex forensic cases by providing a level of discrimination not achievable utilising traditional forensic techniques. The focus of the research was on
Synthesis and characterization of hybrid MCM-41 materials for heavy metal adsorption.
Dimos K, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 126(1), 65-71 (2009)
Mohammad Mahdi Najafpour et al.
Dalton transactions (Cambridge, England : 2003), 41(34), 10156-10160 (2012-07-27)
Nanoscale manganese oxides within Faujasite zeolite have been synthesized with a simple method and characterized by scanning electron microscopy, X-ray diffraction spectrometry, N(2) adsorption-desorption isotherms, transmission electron microscopy, and atomic absorption spectroscopy. These oxides showed efficient water oxidizing activity in
Deborah Wakeham et al.
Physical chemistry chemical physics : PCCP, 14(15), 5106-5114 (2012-01-31)
The nanostructure of the ethanolammonium nitrate (EtAN)-air surface has been investigated using X-ray reflectometry (XRR), vibrational sum frequency spectroscopy (VSFS) and neutral impact collision ion scattering spectroscopy (NICISS). The XRR data decays more rapidly than expected for a perfectly sharp
Sari Palmroth et al.
Annals of botany, 111(3), 467-477 (2013-01-10)
Water and nitrogen (N) are two limiting resources for biomass production of terrestrial vegetation. Water losses in transpiration (E) can be decreased by reducing leaf stomatal conductance (g(s)) at the expense of lowering CO(2) uptake (A), resulting in increased water-use

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service