Skip to Content
Merck
All Photos(1)

Key Documents

750840

Sigma-Aldrich

Ammonium nitrate-18O3

95 atom % 18O, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4N18O3
Molecular Weight:
86.04
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

95 atom % 18O

Assay

98% (CP)

form

solid

bp

210 °C (lit.)

mp

169 °C (lit.)

mass shift

M+6

SMILES string

[18O-][N+]([18O-])=[18O].[NH4+]

InChI

1S/NO3.H3N/c2-1(3)4;/h;1H3/q-1;/p+1/i2+2,3+2,4+2;

InChI key

DVARTQFDIMZBAA-HHKSVJRBSA-O

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1C - Ammonium nitrate and ammonium nitrate containing preparations

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service