Skip to Content
Merck
All Photos(1)

Key Documents

80177

Supelco

Micro particle size standard based on polystyrene monodisperse

analytical standard, size: 2.0 μm

Synonym(s):

Monodisperse polystyrene latex particles (2 μm), Size standards

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€270.00

€270.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
€270.00

About This Item

MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€270.00


Estimated to ship on25 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

form

aqueous suspension
particles

crosslinking

0 % cross-linked

concentration

2% (solids)

particle size

2.0 μm std dev <0.05 μm, coeff var <3%

application(s)

glass & ceramic
industrial qc
pharmaceutical

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Polystyrene-based monodisperse microparticles (size: 2 μm) are a particle size reference standard, ideal for determining the particle size distribution (PSD) profile of test samples.

Application

Used to calibrate, validate and monitor the performance of particle sizers, surface scanning equipment and particle counters.
Monodisperse polystyrene latex particles are also used to investigate their effect on laser-induced breakdown spectroscopy (LIBS) emissions.

Features and Benefits

  • Suitable for routine instrument calibration checks and corrections
  • Available in 5 mL pack size as a neat sample

Analysis Note

For every lot exact values of particle size and standard deviation are determined with an accuracy of 0.001 μm. The size determination of these particles follows procedures described by National Institute of Standards and Technology (NIST, USA) respectively by the Community Bureau of Reference (BCR), esp.:
  • Transmission electron microscopy (TEM)
  • Scanning electron microscope
  • Light microscopy
  • Coulter counter with MDF-system
Calibration of equipment is carried out with NIST-respectively BCR-particle size standards.

Legal Information

Coulter is a trademark of Beckman Coulter, Inc.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

410.0 °F

Flash Point(C)

210 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emmanuel Boateng et al.
Life sciences, 251, 117634-117634 (2020-04-07)
Neuregulin-1β (NRG-1) is a membrane-bound or secreted growth and differentiation factor that mediates its action by binding to ErbB receptors. Circulating levels of NRG-1 are characterized by large inter-individual variability with the range of absolute values covering two orders of
High ErbB3 activating activity in human blood is not due to circulating neuregulin-1 beta
Boateng E, et al.
Life Sciences (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service