Skip to Content
Merck
All Photos(1)

Key Documents

78437

Supelco

Palladium Standard for AAS

TraceCERT®, 1 g/L Pd in hydrochloric acid (nominal concentration)

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€56.10

€56.10


Estimated to ship on08 April 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€56.10

About This Item

UNSPSC Code:
12141733
eCl@ss:
38191001
NACRES:
NA.24

€56.10


Estimated to ship on08 April 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

concentration

1 g/L Pd in hydrochloric acid (nominal concentration)

technique(s)

AAS: suitable

application(s)

industrial qc
pharmaceutical

format

single component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

Palladium Standard for AAS can be used as a standard to determine the amount of palladium biosorbed by Chlorella vulgaris and Saccharomyces cerevisiae immobilized on silica gel from wastewater samples using graphite atomic absorption spectrophotometry (GFAAS).[1]

Analysis Note

1 g/L Pd in 5% hydrochloric acid (contains HNO3 traces), prepared with high purity Pd metal, HCl, HNO3 and water

Other Notes

For a complete product listing of our TraceCERT® range of CRMs for ICP and AAS, technical information and example certificates, please visit our ICP & AAS standards website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Biosorption of platinum and palladium for their separation/preconcentration prior to graphite furnace atomic absorption spectrometric determination
Godlewska-Zylkiewicz B
Spectrochimica Acta. Part B: Atomic Spectroscopy, 58, 1531-1540 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service