Skip to Content
Merck
All Photos(1)

Key Documents

625718

Millipore

Triethylammonium Acetate, 1 M Solution

Ready-to-use buffer solution used for the purification of chemically synthesized oligonucleotides by HPLC. Has a pH of 7.0.

Synonym(s):

Triethylammonium Acetate, 1 M Solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€420.00

€420.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 L
€420.00

About This Item

Empirical Formula (Hill Notation):
C8H19NO2
CAS Number:
Molecular Weight:
161.24
MDL number:
UNSPSC Code:
12161706
NACRES:
NA.25

€420.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

form

solution

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

evapn. residue

<0.01%

color

clear

shipped in

ambient

storage temp.

15-25°C

SMILES string

[N+H](CC)(CC)CC.[O-]C(=O)C

InChI

1S/C6H15N.C2H4O2/c1-4-7(5-2)6-3;1-2(3)4/h4-6H2,1-3H3;1H3,(H,3,4)

InChI key

AVBGNFCMKJOFIN-UHFFFAOYSA-N

General description

Triethylammonium acetate is a mixture of triethylamine (TEA) and acetic acid (AcOH). This is generally used as a solvent in organic synthesis. Triethylammonium acetate is known as a protic ionic liquid since it is a combination of Bronsted acid and base.

Application

Triethylammonium Acetate, 1 M Solution - CAS 5204-74-0 – calbiochem has been used:
  • as a component of high-performance liquid chromatography (HPLC) buffer D and E for the “SMCC-TriNTA purification” program
  • as a component of HPLC buffer A for the purification of commercial 5-hydroxymethyl dCTP (5-hmdCTP) using HPLC technique
  • along with acetonitrile as a buffering reagent for the purification of SMCC-linked TriNTA by reverse-phase HPLC (RP-HPLC)

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miao Yu et al.
Nature protocols, 7(12), 2159-2170 (2012-12-01)
A complete understanding of the potential function of 5-hydroxymethylcytosine (5-hmC), a DNA cytosine modification in mammalian cells, requires an accurate single-base resolution sequencing method. Here we describe a modified bisulfite-sequencing method, Tet-assisted bisulfite sequencing (TAB-seq), which can identify 5-hmC at
Enrico Bodo
Molecules (Basel, Switzerland), 25(6) (2020-04-05)
I have explored the structural features and the dynamics of triethylammonium acetate by means of semi-empirical (density functional tight binding, DFTB) molecular dynamics. I find that the results from the present simulations agree with recent experimental determinations with only few

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service