75113
4-Aminophenyl phosphate monosodium salt hydrate
for electrochemical analysis, ≥98.0% (TLC)
Synonym(s):
p-Aminophenyl phosphate monosodium salt hydrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
for electrochemical analysis
Quality Level
Assay
≥98.0% (TLC)
form
powder
impurities
≤30% water
storage temp.
−20°C
SMILES string
NC1=CC=C(OP(O)(O[Na])=O)C=C1
InChI
1S/C6H8NO4P.Na.H2O/c7-5-1-3-6(4-2-5)11-12(8,9)10;;/h1-4H,7H2,(H2,8,9,10);;1H2/q;+1;/p-1
InChI key
XWJZNVNMMLRLJR-UHFFFAOYSA-M
Application
4-Aminophenyl phosphate monosodium salt hydrate may be used as a reagent for the sensitive detection and estimation of tumor necrosis factor-alpha (TNF-α) in undiluted serum developed via the process of electrochemical enzyme-linked immunosorbent assay (ELISA).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Electrochemical immunosensor for tumor necrosis factor-alpha detection in undiluted serum.
Methods, 116, 125-131 (2017)
Analytical chemistry, 84(2), 1049-1055 (2012-01-03)
This paper reports chemical-chemical (CC) and electrochemical-chemical-chemical (ECC) redox cycling, for use in ultrasensitive biosensor applications. A triple chemical amplification approach using an enzymatic reaction, CC redox cycling, and ECC redox cycling is applied toward electrochemical immunosensors of cardiac troponin
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service