Skip to Content
Merck
All Photos(3)

Key Documents

A5898

Sigma-Aldrich

4-Aminobenzylphosphonic acid

≥95%

Synonym(s):

(4-Aminobenzyl)phosphonic acid, P-[(4-Aminophenyl)methyl]phosphonic acid, [(4-Aminophenyl)methyl]phosphonic acid, p-Aminobenzylphosphonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€83.20
1 G
€313.00

€83.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€83.20
1 G
€313.00

About This Item

Empirical Formula (Hill Notation):
C7H10NO3P
CAS Number:
Molecular Weight:
187.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€83.20


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

powder

storage temp.

−20°C

SMILES string

Nc1ccc(CP(O)(O)=O)cc1

InChI

1S/C7H10NO3P/c8-7-3-1-6(2-4-7)5-12(9,10)11/h1-4H,5,8H2,(H2,9,10,11)

InChI key

NEKHKXMBGWNTOO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service