Skip to Content
Merck
All Photos(1)

Documents

75073

Supelco

Sodium octyl sulfate

suitable for ion pair chromatography, LiChropur, ≥99.0% (T)

Synonym(s):

Octyl sulfate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7OSO3Na
CAS Number:
Molecular Weight:
232.27
Beilstein:
3658635
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21

description

anionic

Quality Level

Assay

≥99.0% (T)

form

crystals

quality

LiChropur

mol wt

232.27 g/mol

technique(s)

ion pair chromatography: suitable

mp

195 °C (dec.) (lit.)

λ

10 % in H2O

UV absorption

λ: 210 nm Amax: 0.1
λ: 220 nm Amax: 0.06
λ: 230 nm Amax: 0.04
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for RP gradient test
corresponds to standard for filter test

SMILES string

[Na+].CCCCCCCCOS([O-])(=O)=O

InChI

1S/C8H18O4S.Na/c1-2-3-4-5-6-7-8-12-13(9,10)11;/h2-8H2,1H3,(H,9,10,11);/q;+1/p-1

InChI key

WFRKJMRGXGWHBM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium octyl sulfate (SOS) is an ion-pair reagent, an anionic surfactant with an 8-carbon tail suitable for cationic separation sorted by carbon chain length.

Application

Sodium octyl sulfate (SOS) may be used as an ion-pairing reagent:
  • For the retention of catechols in reversed-phase high-performance liquid chromatography of catecholamines and their congeners.
  • For the separation of asymmetric dimethylhydrazine (ADMH) and its decomposition products using ion-pair chromatography.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of 1, 1-dimethylhydrazine and its decomposition products using ion-pair chromatography
Ponomarenko SA, et al.
Moscow University Chemistry Bulletin, 64(3), 147-153 (2009)
Reversed-phase high performance liquid chromatography of catecholamines and their congeners with simple acids as ion-paring reagents
Asmus PA and Freed CR
Journal of Chromatography A, 169, 303-311 (1979)
Electrostatic effects on the phase behavior of aqueous cetyltrimethylammonium bromide and sodium octyl sulfate mixtures with added sodium bromide
Brasher LL, et al.
Langmuir, 11(11), 4267-4277 (1995)
Eliseo A Eugenin et al.
Journal of neurochemistry, 151(5), 558-569 (2019-08-06)
Chemical and electrical synapses are the two major communication systems that permit cell-to-cell communication within the nervous system. Although most studies are focused on chemical synapses (glutamate, γ-aminobutyric acid, and other neurotransmitters), clearly both types of synapses interact and cooperate
Miriam Gochin et al.
Journal of medicinal chemistry, 52(14), 4338-4344 (2009-06-19)
The hydrophobic pocket within the coiled coil domain of HIV-1 gp41 is considered to be a hot-spot suitable for small molecule intervention of fusion, although so far it has yielded only microM inhibitors. Previous peptide studies have identified specific hydrophobic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service