Skip to Content
Merck
All Photos(2)

Key Documents

74701

Supelco

Methyl octacosanoate

analytical standard

Synonym(s):

Octacosanoic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€114.00

€114.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
250 MG
€114.00

About This Item

Linear Formula:
CH3(CH2)26COOCH3
CAS Number:
Molecular Weight:
438.77
Beilstein:
1802980
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€114.00


Estimated to ship on24 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

67-69 °C

format

neat

functional group

ester

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C29H58O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28-29(30)31-2/h3-28H2,1-2H3

InChI key

ZKHOYAKAFALNQD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Garlito et al.
Analytica chimica acta, 1051, 103-109 (2019-01-22)
Gas chromatography coupled to a quadrupole time-of-flight mass analyzer (QTOF) with an atmospheric pressure chemical ionization (APCI) source has been tested to study the ionization and mass spectrometric behavior of long-chain and very long-chain polyunsaturated fatty acids (LC-PUFAs, C18-24; VLC-PUFAs
Mónica Narváez-Rivas et al.
Analytica chimica acta, 954, 140-150 (2017-01-14)
The confident identification and in-depth profiling of molecular lipid species remain to be a challenge in lipidomics analysis. In this work, an off-line two-dimensional mixed-mode and reversed-phase liquid chromatography (RPLC) method combined with high-field quadrupole orbitrap mass spectrometer (Q Exactive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service