Skip to Content
Merck
Methyl octacosanoate analytical standard

Methyl octacosanoate

Methyl octacosanoate analytical standard
Synonym(s):
Octacosanoic acid methyl ester
Linear Formula:
CH3(CH2)26COOCH3
CAS Number:
Molecular Weight:
438.77
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Functional Group
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing