Skip to Content
Merck
All Photos(2)

Key Documents

16383

Supelco

Sennoside D

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€1,190.00

€1,190.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€1,190.00

About This Item

Empirical Formula (Hill Notation):
C42H40O19
CAS Number:
Molecular Weight:
848.76
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

€1,190.00


Estimated to ship on24 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](Oc2cccc3[C@@H]([C@H]4c5cccc(O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)c5C(=O)c7c(O)cc(cc47)C(O)=O)c8cc(CO)cc(O)c8C(=O)c23)[C@H](O)[C@@H](O)[C@@H]1O

Related Categories

General description

Sennoside D is an active constituent of senna which is generally used as a laxative.[1] It belongs to anthranoid family, and is procured from dried leaflets and pods of senna plant.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Review article: anthranoid laxatives and their potential carcinogenic effects.
van Gorkom BA
Alimentary Pharmacology & Therapeutics, 13(4), 443-452 (1999)
K Yamauchi et al.
The Journal of pharmacy and pharmacology, 44(12), 973-976 (1992-12-01)
Sennosides A and C directly injected into the caecum of mice showed equal purgative activity. Intracaecal administration reduced time to onset of diarrhoea induced by sennoside C from about 3 h after oral administration to about 24 min. At 2.3
Analytical studies on the active constituents in crude drugs. IV. Determination of sennosides in senna and formulations by high-performance liquid chromatography.
Hayashi S
Chemical & Pharmaceutical Bulletin, 28(2), 406-412 (1980)
T Yagi et al.
The Journal of pharmacy and pharmacology, 49(1), 22-25 (1997-01-01)
This study aimed to explore the mechanism involved in the synergistic purgative action of aloe-emodin anthrone and rhein anthrone, the active metabolites of sennoside C. Aloe-emodin anthrone and rhein anthrone, and their equimolar mixture, induced excretion of an approximately equal
Potentiating effect of sennoside C on purgative activity of sennoside A in mice.
K Kisa et al.
Planta medica, 42(3), 302-303 (1981-07-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service