Skip to Content
Merck
All Photos(1)

Key Documents

73124

Supelco

Bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether

analytical standard

Synonym(s):

2-[4-(3-Chloro-2-hydroxypropyloxy)pheny]-2-[4-(glycidyloxy)phenyl]propane, BADGE-HCl

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€102.00
100 MG
€329.00

€102.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€102.00
100 MG
€329.00

About This Item

Empirical Formula (Hill Notation):
C21H25ClO4
CAS Number:
Molecular Weight:
376.87
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€102.00


Estimated to ship on22 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥90% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

CC(C)(c1ccc(OCC(O)CCl)cc1)c2ccc(OCC3CO3)cc2

InChI

1S/C21H25ClO4/c1-21(2,15-3-7-18(8-4-15)24-12-17(23)11-22)16-5-9-19(10-6-16)25-13-20-14-26-20/h3-10,17,20,23H,11-14H2,1-2H3

InChI key

HLLOKZYCSSQYEB-UHFFFAOYSA-N

General description

Bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether is a chlorinated derivative, which is released during the thermal coating treatment of food cans, big storage vessels and food containers.[1]

Application

Bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether may be used as a reference standard for the determination of bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether in canned food and beverages using fast liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Standard for determination of toxic BADGE and derivatives in canned food[2][3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Biedermann et al.
Mitteilungen Aus Dem Gebiete Der Lebensmittel-Untersuchung Un Hygiene = Travaux De Chimie Alimentaire Et D'Hygiene, 90, 177-177 (1999)
Ans Punt et al.
Archives of toxicology, 93(7), 1941-1953 (2019-05-22)
The goal of the present study was to obtain an in vivo relevant prioritization method for the endocrine potencies of different polycarbonate monomers, by combining in vitro bioassay data with physiologically based kinetic (PBK) modelling. PBK models were developed for
J E Biles et al.
Journal of agricultural and food chemistry, 47(5), 1965-1969 (1999-12-20)
Migration of the diglycidyl ether of bisphenol A (DGEBA) to food from can enamels and can pull-top seals is reported. Derivatives of DGEBA are also determined in some foods. Levels of DGEBA in the foods surveyed in this study range
H Gallart-Ayala et al.
Journal of chromatography. A, 1218(12), 1603-1610 (2011-02-22)
In this work a fast liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS) method using a C18 Fused Core™ column, was developed for the simultaneous analysis of bisphenol A diglycidyl ether (BADGE), bisphenol A (2,3-dihydroxypropyl) glycidyl ether (BADGE·H(2)O), bisphenol A
Antía Lestido Cardama et al.
Polymers, 11(12) (2019-12-19)
The coatings used in cans can release complex chemical mixtures into foodstuffs. Therefore, it is important to develop analytical methods for the identification of these potential migrant compounds in packaged food to guarantee the compliance with European food packaging legislation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service