Skip to Content
Merck
All Photos(1)

Key Documents

61711

Supelco

Bisphenol F bis(2,3-dihydroxypropyl) ether

para-para isomer, analytical standard

Synonym(s):

Bis[4-(2,3-dihydroxypropoxy)phenyl]methane

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€150.00

€150.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€150.00

About This Item

Empirical Formula (Hill Notation):
C19H24O6
CAS Number:
Molecular Weight:
348.39
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€150.00


Estimated to ship on25 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

OCC(O)COc1ccc(Cc2ccc(OCC(O)CO)cc2)cc1

InChI

1S/C19H24O6/c20-10-16(22)12-24-18-5-1-14(2-6-18)9-15-3-7-19(8-4-15)25-13-17(23)11-21/h1-8,16-17,20-23H,9-13H2

InChI key

VGJRHUPDMVWBFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bisphenol F bis(2,3-dihydroxypropyl) ether is the hydrolyzed derivative of bisphenol F-diglycidyl ether (BFDGE), which is produced, when the internal coating of the food storage-can, containing BFDGE comes into contact with aqueous and acidic foodstuffs.[1]

Application

Bisphenol F bis(2,3-dihydroxypropyl) ether may be used as an analytical reference standard for the quantification of the analyte in canned food and beverages,[1] and water-based food stimulants[2] using chromatography techniques.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation of bisphenol A diglycidyl ether, bisphenol F diglycidyl ether and their hydroxy and chlorohydroxy derivatives stability in water-based food simulants
Poustkova I, et al.
Czech Journal of Food Sciences, 22, 272-272 (2004)
H Gallart-Ayala et al.
Journal of chromatography. A, 1218(12), 1603-1610 (2011-02-22)
In this work a fast liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS) method using a C18 Fused Core™ column, was developed for the simultaneous analysis of bisphenol A diglycidyl ether (BADGE), bisphenol A (2,3-dihydroxypropyl) glycidyl ether (BADGE·H(2)O), bisphenol A

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service