Skip to Content
Merck
All Photos(1)

Key Documents

72190

Supelco

Nessler′s reagent

for qualitative det. of ammonia and ammonium compounds

Synonym(s):

Potassium tetraiodomercurate – Potassium hydroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41113319
PubChem Substance ID:
NACRES:
NA.21
form:
liquid

form

liquid

Quality Level

quality

for qualitative det. of ammonia and ammonium compounds

specific analyte(s)

ammonium

technique(s)

photometry: suitable

impurities

potassium hydroxide
potassium tetraiodomercurate

density

1.16 g/mL at 20 °C

application(s)

environmental
food and beverages
general analytical

SMILES string

[K+].[K+].I[Hg--](I)(I)I

InChI

1S/Hg.4HI.2K/h;4*1H;;/q+2;;;;;2*+1/p-4

InChI key

OPCMAZHMYZRPID-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Nessler′s reagent is widely used in determination of ammonium compounds. It can be prepared using anhydrous mercuric iodide and anhydrous potassium iodide. It can be prepared from potassium tetraiodomercurate dissolved in potassium hydroxide.[1]

Application

Nessler′s reagent has been used in determining ammonia in flow analysis regime using analytical spectrometry.[1]

Caution

may form little sediment upon storage

Other Notes

Reagent for the determination of ammonia[2]
The method detection limit is 0.1 mg/L.
Reference: APHA 1975.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT RE 2

Target Organs

Kidney,Central nervous system

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A novel optoelectronic detector and improved flow analysis procedure for ammonia determination with Nessler?s reagent.
Kolacinska, Kamila, and Robert Koncki.
Analytical Sciences, 30.10, 1019-1022 (2014)
E.A. Burns et al.
The Analytical Chemistry of Nitrogen and its compounds, 51-51 (1970)
S J Lambert et al.
Acta crystallographica. Section D, Biological crystallography, 55(Pt 5), 1048-1051 (1999-04-27)
Intact histone octamers, produced by a new method quickly and in bulk, were crystallized in KCl/phosphate, and the X-ray data were analysed to 2.15 A, confirming a P65 space group. This environment preserves the high-resolution structure of the octamers and
Justyna Bzura et al.
Enzyme and microbial technology, 123, 1-7 (2019-01-29)
Two fully mechanized flow analysis systems for urease activity assays have been developed, characterized and compared. Both of them are based on almost the same compact system of solenoid micropumps and microvalves controlled and actuated by highly effective, low-power and
Santosh Kumar Verma et al.
Journal of AOAC International, 90(4), 1180-1190 (2007-09-01)
This paper presents development of a simple, rapid, and precise analytical method for determination of sulfur dioxide in ambient air by a gas to solid-phase conversion method. Sulfur dioxide is determined in the form of sulfite (SO3(2-)) because the absorbing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service