Skip to Content
Merck

Skip To

1.09028

Nessler′s reagent

Synonym(s):

Nota 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mL
Check Cart for Availability
€ 35,50
500 mL
Check Cart for Availability
€ 114,00

About This Item

NACRES:
NB.21
UNSPSC Code:
12352402

€ 35,50


Check Cart for Availability

Request a Bulk OrderRequest more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

specific analyte(s)

nitrogen

technique(s)

photometry: suitable

density

1.16 g/cm3 at 20 °C

General description

for ammonium salts

Preparation Note

Storage temperature: no restrictions.

Analysis Note

Application test:conforms

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.09011C-43080GERPN3004
specific analyte(s)

nitrogen

specific analyte(s)

nitrogen

specific analyte(s)

-

specific analyte(s)

-

technique(s)

photometry: suitable

technique(s)

photometry: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

-

density

1.16 g/cm3 at 20 °C

density

1.25 g/cm3 at 20 °C

density

-

density

-

form

liquid

form

liquid

form

aqueous solution

form

-

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

-

microbialanalysis
microbialanalysis

microbialanalysis
microbialanalysis

-

-


signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT RE 2

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

target_organs

Kidney,Central nervous system

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

7783-33-7

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters