Skip to Content
Merck

Skip To

68367

Prochloraz-(ethylene-d4)

PESTANAL®, analytical standard

Synonym(s):

N-Propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl-d4]-1H-imidazole-1-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
€ 334,00

About This Item

Empirical Formula (Hill Notation):
C15D4H12Cl3N3O2
Molecular Weight:
380.69
MDL number:
UNSPSC Code:
77101502
NACRES:
NA.24

€ 334,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

isotopic purity

≥95.0 atom % D

product line

PESTANAL®

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

SMILES string

O=C(N1C=NC=C1)N(CCC)C([2H])([2H])C([2H])([2H])OC2=C(Cl)C=C(Cl)C=C2Cl

InChI

1S/C15H16Cl3N3O2/c1-2-4-20(15(22)21-5-3-19-10-21)6-7-23-14-12(17)8-11(16)9-13(14)18/h3,5,8-10H,2,4,6-7H2,1H3/i6D2,7D2

InChI key

TVLSRXXIMLFWEO-KXGHAPEVSA-N

General description

Prochloraz-(ethylene-d4) is an isotope-labeled analog of the imidazole fungicide, prochloraz.

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The main advantage of using the IDMS technique is because isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analog, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
198473450834507
format

neat

format

neat

format

neat

format

neat

assay

≥98.0% (HPLC)

assay

≥98.0% (HPLC)

assay

-

assay

-

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

product line

PESTANAL®

product line

PESTANAL®

product line

PESTANAL®

product line

PESTANAL®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

isotopic purity

≥95.0 atom % D

isotopic purity

-

isotopic purity

-

isotopic purity

-


pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters