Skip to Content
Merck
All Photos(1)

Key Documents

34508

Supelco

Prochloraz Metabolite BTS40348

PESTANAL®, analytical standard

Synonym(s):

N-[2-(2,4,6-Trichlorophenoxy)ethyl]propylamine, N-Propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]amine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€386.00

€386.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€386.00

About This Item

Empirical Formula (Hill Notation):
C11H14Cl3NO
CAS Number:
Molecular Weight:
282.59
Beilstein:
6401835
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

€386.00


Estimated to ship on19 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

ClC1=C(OCCNCCC)C(Cl)=CC(Cl)=C1

InChI

1S/C11H14Cl3NO/c1-2-3-15-4-5-16-11-9(13)6-8(12)7-10(11)14/h6-7,15H,2-5H2,1H3

InChI key

CLFQSOIBYICELN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service