Skip to Content
Merck
All Photos(2)

Key Documents

61302

Millipore

Lactalbumin hydrolysate

suitable for microbiology

Synonym(s):

Peptone from lactalbumin

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€124.00

€124.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
€124.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

€124.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

bovine milk

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

total nitrogen (N), ≥12%

ign. residue

≤6%

loss

≤6% loss on drying

pH

6.8-8.0 (25 °C, 2% in H2O)

solubility

H2O: 2%, clear (light yellow to yellow and light brown-yellow to brown-yellow and light brown to brown)

application(s)

food and beverages
microbiology

Looking for similar products? Visit Product Comparison Guide

Application

Lactalbumin contains high levels if essential amino acids. It is recommended for applications requiring a highly soluble source of amino acids and peptides. It is especially useful in fermentations, laboratory media and in tissue culture.

Other Notes

Sales restrictions may apply

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bertha Chen et al.
Fertility and sterility, 91(4 Suppl), 1351-1354 (2008-06-17)
We investigated the effect of relaxin on extracellular matrix remodeling in cultured myometrial compared with leiomyoma smooth muscle cells. Relaxin increases elastase activity and protease inhibitor expressions in myometrial smooth muscle cells, but no effect was observed in leiomyoma cells.
Lesley Bell-Sakyi et al.
Experimental & applied acarology, 49(3), 209-219 (2009-03-03)
Primary cell cultures (n = 16) were initiated from tissues of embryonic and neonatal larval Ornithodoros moubata following methods developed for hard ticks. After maintenance for 20-25 months in vitro, cell multiplication commenced in surviving cultures, leading to the establishment

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service