Skip to Content
Merck

Skip To

61300

M-Gredient® Lactalbumin Peptone

Encymatically Digested, for Biotechnology and Microbiology

Synonym(s):

Animal origin peptone, hydrolyzed protein, Peptone from lactalbumin, Lactalbumin hydrolysate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 g
Check Cart for Availability
€ 147,00
5 kg
Check Cart for Availability
€ 1.240,00

About This Item

CAS Number:
UNSPSC Code:
41106212
NACRES:
NA.74
EC Number:
293-432-6
MDL number:

€ 147,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine milk

Quality Segment

description

Nutrient for fermentations and other applications

form

powder

shelf life

limited shelf life, expiry date on the label

composition

total nitrogen (N), ≥11%

storage condition

dry at room temperature

ign. residue

≤10%

loss

≤6% loss on drying

pH

6.8±0.5 (25 °C, 2% in H2O)

solubility

H2O: 2%, clear to very faintly turbid (light yellow to yellow and light brown-yellow to brown-yellow and light brown to brown)

application(s)

food and beverages
microbiology

General description

Lactalbumin hydrolysate is an enzymatic digest of Lactalbumin and contains high levels of essential amino acids. It is recommended for applications requiring a highly soluble source of amino acids and peptides. It is especially useful in fermentation, laboratory media, and in tissue culture. Lactalbumin, also called whey protein, is the milk albumin, and obtained from whey. There are alpha and beta lactalbumins which are both contained in milk. Owing to its high nutritional quality it can support the growth of numerous cell types like mammalian, insect, and bacterial.

Application

Lactalbumin hydrolysate can be used: -
In the production of recombinant proteins
As a supplement in mammalian cell culture media
As a substrate for growth of many microbes

Other Notes

Sales restrictions may apply

Legal Information

M-GREDIENT is a registered trademark of Merch KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
613028230370173
biological source

bovine milk

biological source

bovine milk

biological source

bovine milk

biological source

bovine milk

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

application(s)

food and beverages, microbiology

application(s)

food and beverages, microbiology

application(s)

food and beverages
microbiology

application(s)

food and beverages, microbiology

form

powder

form

powder

form

powder

form

powder

solubility

H2O: 2%, clear to very faintly turbid (light yellow to yellow and light brown-yellow to brown-yellow and light brown to brown)

solubility

H2O: 2%, clear (light yellow to yellow and light brown-yellow to brown-yellow and light brown to brown)

solubility

H2O: 2%, clear, yellow to tan, water: soluble 2%, clear, faintly brown-yellow

solubility

H2O: 2%, clear, yellow (and faint yellow and faint brown)

pH

6.8±0.5 (25 °C, 2% in H2O)

pH

6.8-8.0 (25 °C, 2% in H2O)

pH

7.0±0.5 (2% in H2O)

pH

6.5-7.5 (2% in H2O)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters