Skip to Content
Merck
All Photos(1)

Key Documents

59799

Supelco

4-(Diethylaminomethyl)benzhydrazide

for HPLC derivatization, LiChropur, ≥98.0% (HPLC)

Synonym(s):

4-(Diethylaminomethyl)benzoic acid hydrazide, 4-(Diethylaminomethyl)benzoic hydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19N3O
CAS Number:
Molecular Weight:
221.30
Beilstein:
13743368
MDL number:
UNSPSC Code:
41121800
PubChem Substance ID:

grade

for HPLC derivatization

Quality Level

Assay

≥98.0% (HPLC)

quality

LiChropur

storage temp.

2-8°C

SMILES string

O=C(NN)C1=CC=C(CN(CC)CC)C=C1

InChI

1S/C12H19N3O/c1-3-15(4-2)9-10-5-7-11(8-6-10)12(16)14-13/h5-8H,3-4,9,13H2,1-2H3,(H,14,16)

InChI key

GIGSNSOHCRCJJS-UHFFFAOYSA-N

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshio Suzuki et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 20(3), 475-482 (2004-04-08)
Novel labeling reagents, called MS probes, which possess a positively charged quaternary amine moiety and can transform a neutral analyte into a charged compound by simply mixing with the analyte and allowing the mixture to stand from several minutes to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service