Skip to Content
Merck
All Photos(1)

Key Documents

913642

Sigma-Aldrich

Silica nanoparticles

pore size non-porous, 150 nm particle size, 5 % (w/w) in ethanol, dispersion

Synonym(s):

Monodisperse silicon dioxide, Non-porous silica, Non-porous silicon dioxide, Silica

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
€188.00

€188.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 ML
€188.00

About This Item

Empirical Formula (Hill Notation):
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
12352309
NACRES:
NA.23

€188.00


Check Cart for Availability

Request a Bulk Order

form

dispersion
nanoparticles

Quality Level

concentration

5 % (w/w) in ethanol

particle size

150 nm

pore size

non-porous

SMILES string

[Si](=O)=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Monodisperse nonporous silica nanoparticles find application in drug delivery and molecular imaging[1][2] whereas micron sized particles are used in liquid chromatography[3]. We offer highly monodisperse nonporous silica in ethanol, with sizes varying from 150 nm to 900 nm. The high monodispersity of our particles makes them suitable for aforementioned applications.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Tang et al.
Nano today, 8(3), 290-312 (2013-09-03)
Nanomedicine, the use of nanotechnology for biomedical applications, has potential to change the landscape of the diagnosis and therapy of many diseases. In the past several decades, the advancement in nanotechnology and material science has resulted in a large number

Articles

Nonporous SiNPs are investigated as nanocarriers for drug delivery and smart plastics due to monodispersed particles.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service