Skip to Content
Merck
All Photos(2)

Key Documents

59744

Millipore

Hybriscan D kit

Lactobac, suitable for food and beverages, microbiology and in situ hybriziation

Synonym(s):

FastScan Lactobac, Lactobacilli HybriScanD

Sign Into View Organizational & Contract Pricing

Select a Size

96 TESTS
€413.00

€413.00


Usually ships in 2 weeks.

Request more information

Select a Size

Change View
96 TESTS
€413.00

About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

€413.00


Usually ships in 2 weeks.

Request more information

Product Name

HybriScanD Lactobac, suitable for microbiology

usage

sufficient for 96 reactions

manufacturer/tradename

(HybriScan)

technique(s)

microbe id | in situ hybriziation: suitable

application(s)

food and beverages
microbiology

storage temp.

2-8°C

suitability

Lactobacillus spp.

General description

HybriScan D kits detect and simultaneously identify microorganisms using rRNA as the detection target. The principle is based on in situ sandwich hybridization with two specific probes. As the rRNA of dead cells is degraded in the presence of RNase, only living cells are detected. Each cell has several thousand rRNA copies, so neither PCR nor a polymerase is needed for this technology, which means that this molecular biology test is not inhibited by the sample matrix, for example food, beverages and water. We recommend a 24-hour enrichment in MRS Broth (Cat. No. 1.10661), Tomato Juice Broth (Cat. No. 17218)

Features and Benefits

Sensitivity (after pre enrichment): 1-10 cfu/L

Legal Information

HybriScan is a trademark of ScanBec GmbH

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

HybriScan™ assays provide rapid, accurate molecular detection of beer spoilage microorganisms, ensuring excellent product quality in breweries.

Bifidobacteria-selective media detects beneficial bacteria, aiding quality control in microbiology labs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service