17154
Lactobacillus bulgaricus Agar (Base)
NutriSelect® Plus, suitable for microbiology
Select a Size
About This Item
Recommended Products
sterility
non-sterile
Quality Level
form
powder
shelf life
limited shelf life, expiry date on the label
composition
agar, 20 g/L
beef extract, 10 g/L
casein enzymic hydrolysate, 10 g/L
dextrose, 20 g/L
dipotassium phosphate, 2 g/L
polysorbate 80, 1 g/L
tomato juice, 2 g/L
yeast extract, 5 g/L
manufacturer/tradename
NutriSelect® Plus
technique(s)
microbiological culture: suitable
final pH
6.8±0.2 (25 °C)
application(s)
environmental
food and beverages
microbiology
suitability
selective by low pH for Lactobacillus spp.
Related Categories
Application
Preparation Note
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Cost-effective molecular biology method for lactobacilli detection is presented as an alternative to PCR-based methods.
Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service