Skip to Content
Merck
All Photos(1)

Key Documents

49861

Supelco

o-Hydroxyhippuric acid

analytical standard

Synonym(s):

2-Hydroxybenzoylaminoacetic acid, Salicylglycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO4
CAS Number:
Molecular Weight:
195.17
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

mp

167-169 °C (lit.)

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)CNC(=O)c1ccccc1O

InChI

1S/C9H9NO4/c11-7-4-2-1-3-6(7)9(14)10-5-8(12)13/h1-4,11H,5H2,(H,10,14)(H,12,13)

InChI key

ONJSZLXSECQROL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Monika Untergehrer et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 57, 396-402 (2019-03-09)
Willow bark (Salicis cortex) is a herbal medicinal drug used to treat fever and pain, such as headaches and lower back pain. Until now, it has not been fully understood which compounds are responsible for the efficacy of the drug.
Elardus Erasmus et al.
PloS one, 14(5), e0216298-e0216298 (2019-05-11)
Chronic fatigue, in its various manifestations, frequently co-occur with pain, sleep disturbances and depression and is a non-communicable condition which is rapidly becoming endemic worldwide. However, it is handicapped by a lack of objective definitions and diagnostic measures. This has
Serena Lucotti et al.
The Journal of clinical investigation, 129(5), 1845-1862 (2019-03-26)
Because metastasis is associated with the majority of cancer-related deaths, its prevention is a clinical aspiration. Prostanoids are a large family of bioactive lipids derived from the activity of cyclooxygenase-1 (COX-1) and COX-2. Aspirin impairs the biosynthesis of all prostanoids
Giovanna Baron et al.
Biochemical pharmacology, 173, 113726-113726 (2019-11-30)
The aim of this work was to profile, by using an HPLC-MS/MS method, cranberry compounds and metabolites found in human urine after ingestion of a highly standardized cranberry extract (Anthocran®). Two different strategies were adopted for the data analysis: a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service