Skip to Content
Merck
All Photos(1)

Key Documents

46905

Sigma-Aldrich

9-Oxofluorene-4-carboxylic acid

purum, ≥97.0% (T)

Synonym(s):

9-Fluorenone-4-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8O3
CAS Number:
Molecular Weight:
224.21
Beilstein:
2694801
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥97.0% (T)

mp

225-227 °C (lit.)
226-228 °C

SMILES string

OC(=O)c1cccc2C(=O)c3ccccc3-c12

InChI

1S/C14H8O3/c15-13-9-5-2-1-4-8(9)12-10(13)6-3-7-11(12)14(16)17/h1-7H,(H,16,17)

InChI key

AFQYQSWTVCNJQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal structure of 9-Oxofluorene-4-carboxylic acid.
Kennard CHL, et al.
Australian Journal of Chemistry, 34(5), 1143-1146 (1981)
613. 4, 10-Diazapyrenes.
Coffin B and Robbins RF.
Journal of the Chemical Society, 3379-3382 (1965)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service