40495-U
cis/trans F.A.M.E Column Performance Mix
2.5 mg/mL in dichloromethane, ampule of 1 mL
About This Item
Recommended Products
form
liquid
CofA
current certificate can be downloaded
packaging
ampule of 1 mL
concentration
2.5 mg/mL in dichloromethane
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
format
multi-component solution
storage temp.
2-8°C
Application
Components
Please note that the components and concentrations may vary from batch to batch.
Typical Composition:
C16:0
C18:0
C18:1 trans isomers
C18:1 cis and trans isomers
C18:1 cis isomers
C18:2
C20:0
C20:1
C18:3
C22:0
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
AOCS and AOAC methods for trans fatty acid composition require highly polar cyanosilicone capillary columns.
Protocols
FAME Standard for Optimizing GC System Performance
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service