Skip to Content
Merck
All Photos(1)

Key Documents

45892

Supelco

Pendimethalin solution

100 μg/mL in methanol, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
€54.70

€54.70


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
2 ML
€54.70

About This Item

Empirical Formula (Hill Notation):
C13H19N3O4
CAS Number:
Molecular Weight:
281.31
Beilstein:
2157711
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€54.70


Estimated to ship on25 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

CCC(CC)Nc1c(cc(C)c(C)c1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C13H19N3O4/c1-5-10(6-2)14-12-11(15(17)18)7-8(3)9(4)13(12)16(19)20/h7,10,14H,5-6H2,1-4H3

InChI key

CHIFOSRWCNZCFN-UHFFFAOYSA-N

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Tsukada et al.
Singapore medical journal, 50(3), e105-e106 (2009-04-09)
The herbicide, pendimethalin, is used worldwide, but its acute toxicity is not yet widely known. There have been some reported acute pendimethalin poisoning cases in humans and most of them intentionally ingested the concentrated formulation. We describe a 73-year-old man
Indu Chopra et al.
Environmental monitoring and assessment, 160(1-4), 123-126 (2009-01-09)
The mobility of pendimethalin in sandy loam soil was studied in soil columns under laboratory conditions at two application rates, 1.0 and 2.0 kg a.i. ha(-1), with simulated rainfall of 300 mm. The maximum concentration of the herbicide was found
U Undeğer et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 48(2), 502-508 (2009-11-10)
The endocrine disrupting potential of the herbicide pendimethalin was investigated in vivo on the uterotrophic response and on the expression of estrogen-regulated genes examined by quantitative real-time RT PCR. Receptor binding characteristics of pendimethalin were analyzed by an in silico
Morgane Danion et al.
Ecotoxicology and environmental safety, 79, 28-34 (2012-03-01)
In this study, the in vivo effects of chronic pollution by the active substance (AS) pendimethalin, a dinitroaniline herbicide, on the susceptibility of rainbow trout, Oncorhynchus mykiss L., to an experimental challenge with viral hemorrhagic septicemia virus (VHSV) were assessed.
Morgane Danion et al.
The Science of the total environment, 424, 143-152 (2012-03-27)
The in vivo effects of the herbicide active substance (AS) pendimethalin (alone and with Prowl 400® adjuvant) were evaluated on sanitary status i.e. the health status with regard to chemical pollution and on the physiological state via the immune system

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service