Skip to Content
Merck
All Photos(1)

Key Documents

45329

Supelco

Asulam

PESTANAL®, analytical standard

Synonym(s):

Methyl N-(4-aminophenylsulfonyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10N2O4S
CAS Number:
Molecular Weight:
230.24
Beilstein:
2697523
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)NS(=O)(=O)c1ccc(N)cc1

InChI

1S/C8H10N2O4S/c1-14-8(11)10-15(12,13)7-4-2-6(9)3-5-7/h2-5H,9H2,1H3,(H,10,11)

InChI key

VGPYEHKOIGNJKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R T Kon et al.
Food additives and contaminants, 1(1), 67-71 (1984-01-01)
An analytical method has been developed for the determination of the herbicide, asulam [methyl(4-aminobenzenesulphonyl)carbamate] and two metabolites, sulphanilamide and acetylasulam individually, in peaches. Asulam and acetylasulam were partitioned from an aqueous phase with ethyl ether, leaving behind more polar components
J K Rowntree et al.
Annals of botany, 92(4), 547-556 (2003-08-23)
Asulox is a herbicide used to control bracken. Its effects on mosses were investigated to ascertain whether exposure proved as detrimental as found in parallel studies on pteridophytes. Mature gametophytes of 18 mosses were exposed to a range of concentrations
Effects of asulam on some microbial activities of three soils.
J A Marsh
Bulletin of environmental contamination and toxicology, 25(1), 15-22 (1980-07-01)
Direct analysis of the wild oat herbicide, asulam, in wheat samples by reversed-phase liquid chromatography at selected ultraviolet wavelengths.
J F Lawrence et al.
Journal of agricultural and food chemistry, 28(6), 1323-1325 (1980-11-01)
Julia A Spoors et al.
Journal of environmental monitoring : JEM, 4(6), 917-921 (2003-01-02)
To date, no ligand binding assay has been described for the carbamate herbicide asulam, although a variety of physical methods, dependent on pre-concentration of water samples, have been documented for its assessment. However, asulam is increasingly used in sensitive agricultural

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service