Skip to Content
Merck
All Photos(2)

Key Documents

32586

Supelco

Florasulam

PESTANAL®, analytical standard

Synonym(s):

N-(2,6-Difluorophenyl)-8-fluoro-5-methoxy-[1,2,4]triazolo[1,5-c]pyrimidine-2-sulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€160.00

€160.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€160.00

About This Item

Empirical Formula (Hill Notation):
C12H8F3N5O3S
CAS Number:
Molecular Weight:
359.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€160.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1ncc(F)c2nc(nn12)S(=O)(=O)Nc3c(F)cccc3F

InChI

1S/C12H8F3N5O3S/c1-23-12-16-5-8(15)10-17-11(18-20(10)12)24(21,22)19-9-6(13)3-2-4-7(9)14/h2-5,19H,1H3

InChI key

QZXATCCPQKOEIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Florasulam is a selective triazolopyrimidine sulfonanilide post-emergent herbicide. Its pesticidal mode of action (MOA) is via inhibition of acetolactate synthase (ALS) in plants. ALS is found in the chloroplast where it catalyses branch chained amino acid biosynthesis. Inhibition of ALS results in inhibition of plant cell division, decreased plant growth, and ultimately, plant death.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service