Skip to Content
Merck
All Photos(1)

Key Documents

44113

Supelco

Cyclohexanol

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€212.00

€212.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
€212.00

About This Item

Linear Formula:
C6H11OH
CAS Number:
Molecular Weight:
100.16
Beilstein:
906744
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€212.00


Estimated to ship on28 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

3.5 (vs air)

vapor pressure

0.98 mmHg ( 25 °C)

Assay

≥98.5% (GC)

autoignition temp.

572 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

12.25 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% cyclohexanone (GC)
≤0.5% water

refractive index

n20/D 1.465 (lit.)

bp

160-161 °C (lit.)

mp

20-22 °C (lit.)

density

0.948 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

OC1CCCCC1

InChI

1S/C6H12O/c7-6-4-2-1-3-5-6/h6-7H,1-5H2

InChI key

HPXRVTGHNJAIIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

147.2 °F - closed cup - DIN 51794

Flash Point(C)

64 °C - closed cup - DIN 51794


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MengTing Zhu et al.
Food chemistry, 321, 126640-126640 (2020-04-06)
The oxidation of cold-pressed and commercial refined camellia oil stored at room temperature for one year was comparatively studied by 1H and 31P NMR spectroscopy. The oxidation of extra virgin olive oil (EVOO) under the same conditions was further compared.
Martin Globisch et al.
Journal of agricultural and food chemistry, 62(33), 8500-8507 (2014-07-30)
Hexanal, 2-heptenal, and nonanal were identified as relevant reaction products formed in the course of the lipid peroxidation of heated peanut oil. For the identification of potential amino acid side chain adducts, kinetic studies between N(α)-benzoylglycyl-l-lysine as a model for
Rui Yang et al.
Analytical and bioanalytical chemistry, 407(3), 737-747 (2014-07-12)
Microfluidic systems with monolithic columns have been developed for preconcentration and on-chip labeling of model proteins. Monoliths were prepared in microchannels by photopolymerization, and their properties were optimized by varying the composition and concentration of the monomers to improve flow
Eric L Kendall et al.
Biomicrofluidics, 8(4), 044109-044109 (2014-11-08)
A technique for microfluidic, pH modulated DNA capture and purification using chitosan functionalized glycidyl methacrylate monoliths is presented. Highly porous polymer monoliths are formed and subsequently functionalized off-chip in a batch process before insertion into thermoplastic microchannels prior to solvent
Roy Bogseth et al.
Journal of the American Society for Mass Spectrometry, 25(11), 1987-1990 (2014-09-25)
Simple alcohols formed protonated acetonitrile adducts containing up to two acetonitrile molecules when analyzed by ESI or APCI in the presence of acetonitrile in the solvent. These acetonitrile adducts underwent dissociation to form a nitrilium ion, also referred to as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service