Skip to Content
Merck
All Photos(1)

Key Documents

8.02824

Sigma-Aldrich

Cyclohexene

(stabilized) for synthesis

Synonym(s):

Cyclohexene, 1,2,3,4-Tetrahydrobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€24.60
500 ML
€30.40
1 L
€39.70
2.5 L
€88.60

€24.60


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€24.60
500 ML
€30.40
1 L
€39.70
2.5 L
€88.60

About This Item

Empirical Formula (Hill Notation):
C6H10
CAS Number:
Molecular Weight:
82.14
MDL number:
UNSPSC Code:
12352002
EC Index Number:
203-807-8
NACRES:
NA.22

€24.60


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

90 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

310 °C

potency

1300 mg/kg LD50, oral (Rat)

expl. lim.

1.2-7.7 % (v/v)

pH

7-8 (20 °C, 0.2 g/L in H2O)

kinematic viscosity

0.82 cSt (calculated)

bp

83 °C/1013 hPa

mp

-104 °C

transition temp

flash point -15.6 °C (DIN 51755 Part 1)

solubility

0.21 g/L

density

0.81 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C1CCC=CC1

InChI

1S/C6H10/c1-2-4-6-5-3-1/h1-2H,3-6H2

InChI key

HGCIXCUEYOPUTN-UHFFFAOYSA-N

Application

  • Preparation of cyclohexene isotopologues and stereoisotopomers from benzene: This groundbreaking study demonstrates the transformation of benzene into various forms of cyclohexene, expanding the potential applications of this compound in scientific research (JA Smith, KB Wilson, RE Sonstrom, PJ Kelleher, 2020).
  • Highly selective oxidation of cyclohexene to 2-cyclohexene-1-one in water using molecular oxygen over Fe–Co–gC3N4: This study explores an environmentally friendly method of oxidizing cyclohexene using molecular oxygen, showing potential for green chemistry applications (D Yang, T Jiang, T Wu, P Zhang, H Han, 2016).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.810 - 0.811
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service