Merck
  • Protic ionic liquids (PILs) nanostructure and physicochemical properties: development of high-throughput methodology for PIL creation and property screens.
Protic ionic liquids (PILs) nanostructure and physicochemical properties: development of high-throughput methodology for PIL creation and property screens.
Physical chemistry chemical physics : PCCP (2014-12-06)
Tamar L Greaves, Krystal Ha, Benjamin W Muir, Shaun C Howard, Asoka Weerawardena, Nigel Kirby, Calum J Drummond
ABSTRACT
A high-throughput approach was developed in order to prepare and dry a series of protic ionic liquids (PILs) from 48 Brønsted acid-base combinations. Many combinations comprised an alkyl carboxylic acid paired with an alkyl amine. Visual screens were developed to identify which acid-base combinations formed PILs, and of those, which PILs were likely to have high surface tensions, low viscosities, and low melting points. The surface tension screen was validated through pendant drop surface tension measurements. Karl Fischer coulometric titration was used to obtain the water contents, and it was noted that there is a considerable difference in the drying rate throughout this series of PILs. It was observed that an octyl chain present on either the cation or anion was detrimental to the formation of a PIL with a low melting point, and instead increased the likelihood of a gel or solid forming. The nanostructure of the PILs was determined, using synchrotron small and wide angle X-ray scattering (SAXS/WAXS), to consist of polar and non-polar domains, with the alkyl chains on the cation and anion intercalating. The results indicate that both the alkyl chain on the cation and/or anion contribute to the correlation distance, for the intermediate range order, with the expectation that there is charge alternation of the ions in the polar region. The maximum correlation distance was observed when there was an alkyl chain present on only one ion. This correlation distance could be significantly reduced by varying the alkyl chain length present on the other ion, which was attributed to increased disorder and interdigitation of chains, and to toe-to-toe alignment of the chains. To the best of our knowledge this is the first PIL report into the effect of having an alkyl chain present on both the cation and the anion.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Amylamine, ≥99%, FG
Sigma-Aldrich
Propylamine, purum, ≥99.0% (GC)
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
Octanoic acid, ≥99%
Supelco
Aucubin, analytical standard
Supelco
Ethanolamine, analytical standard
Sigma-Aldrich
Acetic acid, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
Supelco
Octanoic acid, analytical standard
Sigma-Aldrich
Hexanoic acid, purum, ≥98.0% (GC)
Supelco
Methanol, analytical standard
Supelco
Hexanoic acid, analytical standard
Sigma-Aldrich
Propylamine, 98%
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Amylamines, mixture of isomers, ≥98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Octanoic acid, ≥98%
Sigma-Aldrich
Ethylamine, 97%
Sigma-Aldrich
Octanoic acid, ≥98%, FG
Sigma-Aldrich
Octanoic acid, natural, ≥98%, FG
Sigma-Aldrich
Hexanoic acid, ≥98%, FCC, FG
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Propylamine, ≥99%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Aucubin, primary reference standard
Sigma-Aldrich
Amylamine, 99%