Skip to Content
Merck
All Photos(1)

Key Documents

436658

Supelco

Ambersep 21K XLT Ion Exchange Resin

chloride form, 16-20 mesh

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€185.00

€185.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
€185.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:
NACRES:
SB.52

€185.00


Check Cart for Availability

Request a Bulk Order

Product Name

AMBERSEP® 21K XLT Mesh Anion Exchange Resin (Cl-),

form

beads

Quality Level

crosslinking

<10 % cross-linked

moisture

50-58%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

trimethylamine

particle size

525-625 μm

capacity

1.2 meq/mL by wetted bed volume

separation technique

anion exchange

SMILES string

[Cl-].[N+](C)(C)(C)c3ccc(cc3)C=C.c2(ccc(cc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C11H16N.C10H10.C8H8.ClH/c1-5-10-6-8-11(9-7-10)12(2,3)4;1-3-9-5-7-10(4-2)8-6-9;1-2-8-6-4-3-5-7-8;/h5-9H,1H2,2-4H3;3-8H,1-2H2;2-7H,1H2;1H/q+1;;;/p-1

InChI key

BBQMUEOYPPPODD-UHFFFAOYSA-M

General description

This is mining grade material, and does NOT meet FDA 21 CFR 173.25. For information on the Dowex 21K XLT resin, contact [email protected].

Other Notes

This ion-exchange resin has not been specially processed nor cleaned. We suggest that it be treated with a suitable preliminary elution and wash.

Legal Information

Ambersep is a registered trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Would AmberSep™ 21K Ion Exchange Resin be suitable for recovering palladium from a wastewater stream?

    1 answer
    1. Yes, this is an appropriate resin to use for this application.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service