Skip to Content
Merck
All Photos(1)

Key Documents

217387

Supelco

AmberChrom 1X2 Ion Exchange Resin

chloride form, 100-200 mesh

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€101.00
500 G
€415.00

€101.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
€101.00
500 G
€415.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
SB.52

€101.00


Estimated to ship on22 May 2025


Request a Bulk Order

Product Name

AmberChrom 1X2 chloride form, 100-200 mesh

form

(Powder, Beads or Crystals)

Quality Level

crosslinking

2 % cross-linked

parameter

66 °C OH- form max. temp.
99 °C Cl- form max. temp.

moisture

70-80%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

trimethylbenzylammonium functional group

particle size

100-200 mesh

operating pH

0-14

capacity

0.7 meq/mL by wetted bed volume

separation technique

anion exchange

SMILES string

N(C)(C)C.C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H12.C10H10.C8H8.C3H9N/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8;1-4(2)3/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2;1-3H3

InChI key

HADXLRSCRPYPJJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

AmberChrom 1X2 is an anion exchange resin.[1]

Application

AmberChrom 1X2 chloride form was used to separate cadmium from seawater matrix using column chromatography.[2] It was also used in column for gas chromatography-mass spectrometric analysis.[3]
Strongly basic (type I) anion exchange resin for food & drug processing; meets requirements of FDA Food Additive Regulation 21 CFR 173.25

Features and Benefits

strongly basic anion.

Other Notes

This ion-exchange resin has not been specially processed nor cleaned. We suggest that it be treated to a suitable preliminary elution and wash.

Legal Information

AmberChrom is a trademark or registered trademark of affiliates of DuPont de Nemours, Inc.
Amberchrom is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Association of selected viniviticultural factors with sensory and chemical characteristics of New Zealand Sauvignon blanc wines.
Parr, Wendy V., et al.
Food Research International, 53, 464-475 (2013)
Determination of cadmium in mari waters: on-line preconcentration and flow-through fluorescence detection.
Charles, S., et al.
Analytica Chimica Acta, 440, 37-43 (2001)
Energy transfer-room temperature phosphorescence for the optosensing of transition metal ions.
Di'az-Garci'a, Juan, et al.
Analytica Chimica Acta, 486, 1-10 (2003)

Questions

1–2 of 2 Questions  
  1. How can a Dowex type 1 resin in the chloride form be converted to the carbonate form?

    1 answer
    1. As one might expect, the procedure requires the use of sodium carbonate (Na2CO3).Citation: Journal of Experimental Medicine, 120, 1007-1018 (1965).From page 1011 of that paper:"A 2 mL column of washed Dowex-1X8 200 to 400 mesh, in the chloride form, was converted to the carbonate form by washing with 1 M Na2CO3 until no chloride could be detected in the eluate. It was then washed with distilled water until the pH of the eluate reached neutrality". The simplest test for chloride is to use a dilute silver nitrate solution. If mixing a few drops of the eluate with a few drops of a silver nitrate solution results in a silver chloride precipitate forming, that would an indication that the eluate contains chloride. You could also use a chloride-specific electrode, if you have access to one.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service