Skip to Content
Merck
All Photos(3)

Key Documents

4000BC

Millipore

TRANSIA® AG EHEC Kit

suitable for ELISA testing in Environmental, and Food and Beverages

Sign Into View Organizational & Contract Pricing

Select a Size

96 TESTS
€687.00

€687.00


Estimated to ship on30 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
96 TESTS
€687.00

About This Item

UNSPSC Code:
41116129
NACRES:
NA.85

€687.00


Estimated to ship on30 May 2025


Request a Bulk OrderRequest more information

Product Name

TRANSIA® AG EHEC, BioControl, ELISA test for detection of E. coli O157:H7 in food and environmental samples

Agency

AOAC® (OMA)

product line

TRANSIA®

form

liquid

shelf life

6‑15 mo.

packaging

pkg of 96

manufacturer/tradename

BioControl Systems 4000-01
TRANSIA®

technique(s)

ELISA: suitable

input

food(s)

application(s)

food and beverages
pathogen testing

storage temp.

2-8°C

suitability

Escherichia coli (O157:H7)

General description

TRANSIA AG EHEC is an enzyme immunoassay that detects enterohemorrhagic E. coli. It employs highly specifi c antibodies directed against E. coli O157:H7 antigens and has been specifi cally formulated to minimize cross-reactivity with many Enterobacteriaceae while maintaining superior sensitivity.

Application

AOAC Official Method 996.10

Features and Benefits

  • Accurate detection of E. coli O157:H7 in beef in just 10 hours.Results in 20 hours for other foods.Validated for use with a wide variety of foods including meat, dairy, poultry, fruit, nuts, and more.Ready to use reagents and breakaway wells allow you to use only the number of tests required for your samples, helping to reduce waste and keep testing costs low.

Components

Test wells, wash solution concentrate, substrate, conjugate, positive control, stop solution

Testing Method

ELISA

Legal Information

AOAC is a registered trademark of AOAC International
TRANSIA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

TRANSIA AG (Enzyme ImmunoAssay) and TRANSIA® PLATE incorporate proven antibody-antigen "sandwich" assay technology for the detection of food borne pathogens in both product and environmental samples

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service