Skip to Content
Merck

LI0691BC

TRANSIA® PLATE Listeria spp.

BioControl, ELISA test for detection of Listeria in food and environmental samples

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.85
UNSPSC Code:
41116129
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

AFNOR

product line

TRANSIA®

shelf life

3‑24 mo.

packaging

pkg of 96 tests

manufacturer/tradename

BioControl Systems LI0691

technique(s)

ELISA: suitable

input

food(s)

application(s)

food and beverages
pathogen testing

storage temp.

2-8°C

suitability

Listeria spp.

General description

TRANSIA® PLATE Listeria is a kit for detection of the Listeria genus(L. monocytogenes, L. ivanovii, L. innocua, L. seeligeri, L. welshimeri, L. grayi )in dairy products, meat products, seafood products, vegetables, and environmental samples.

Application

AFNOR Certification TRA 02/06 – 11/95
ELISA

Biochem/physiol Actions

TRANSIA® PLATE Listeria is an ELISA test (Enzyme Linked Immuno Sorbent Assay) based on a two step sandwich-type reaction. The solid support of the reaction is a microtitration plate with dividable microwellstrips. Specific antibodies of flagellar antigens are grafted on the microtitration plate wells.

Features and Benefits

  • Detects all 6 species of Listeria providing a true indication of product and surface quality.
  • Positive and Negative controls supplied with the kit.

Other Notes

  • 1 microplate with dividable microwell strips with 96 wells (12 microwell strips of 8 microwells).
  • Negative control: (bacterial antigens not belonging to the Listeria genus), ready-to-use, 10 mL.
  • Positive control: Listeria monocytogenes antigens, ready-to-use, 8 mL.
  • Conjugate (contains peroxidase conjugated anti-Listeria antibodies), ready-to-use, 16 mL.
  • Wash buffer, 20 X concentration, 60 mL.
  • Substrate (urea H2O2), ready-to-use, 57 mL.
  • Chromogen (TMB), ready-to-use, 57 mL.
  • Stop solution (H2SO4), ready-to-use, 57 mL.
  • 1 microplate cover.

Legal Information

TRANSIA is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library