Skip to Content
Merck
All Photos(1)

Key Documents

39187

Millipore

Universal Environmental ID Membrane

pale pink colored, sterile, diameter 70 mm, suitable for microbiology

Synonym(s):

Indicator membrane, ID Membranes

Sign Into View Organizational & Contract Pricing

Select a Size

25 DISCS
€66.70

€66.70


Estimated to ship on02 June 2025


Request more information

Select a Size

Change View
25 DISCS
€66.70

About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

€66.70


Estimated to ship on02 June 2025


Request more information

membrane size

70 mm

sterility

sterile

shelf life

limited shelf life, expiry date on the label

storage condition

protect from light

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

agriculture
clinical testing
food and beverages
food and beverages

microbiology

storage temp.

2-8°C

suitability

Enterococcus spp.
Escherichia coli
Pseudomonas spp.
Salmonella spp.
coliforms

General description

Universal Environmental ID Membranes are substrate-carrying membranes used for rapid identification and differentiation of environmental pathogens through characteristic color reactions by placing on the agar surface post-incubation. The membranes contain chromogenic substrates such as ONPG, X-Gal, or X-Glu, and other substrates and indicators, which serve as the basis for microbe differentiation by color. The target organisms possess enzyme systems that metabolize the substrates in the membrane, leading to a change of color. The colors can be visually detected on the membrane. This indicates microbial enzymatic activity, which serves to identify the genus and species. This economical, confirmatory, and rapid identification method is suitable for microorganisms from environmental samples, and samples of clinical origin such as nosocomial samples.

Application

Universal Environmental ID Membrane is recommended for rapid detection of Pseudomonas, Enterococcus, E. coli, and Salmonella species, etc., from environmental samples and samples of clinical origin, such as nosocomial samples. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostics, etc.

Analysis Note

Escherichia coli (25922)pink - purpleStaphylococcus aureus (25923) - golden yellowPseudomonas aeruginosa (27853) - colorless (greenish pigment is observed)Enterococcus faecalis (29212) blue – blue green (small)Salmonella Typhimurium (14028) - colorless

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service