Skip to Content
Merck
All Photos(1)

Documents

03719

Millipore

Dual Confirmation of E. coli ID Membrane

white colored, sterile, diameter 70 mm, suitable for microbiology

Synonym(s):

Indicator membrane, E. coli ID Membranes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

membrane size

70 mm

sterility

sterile

shelf life

limited shelf life, expiry date on the label

storage condition

protect from light

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

clinical testing
environmental
food and beverages
pharmaceutical

microbiology

storage temp.

2-8°C

suitability

Escherichia coli
coliforms

General description

Dual Confirmation of E. coli ID Membranes are substrate-carrying membranes used for rapid identification and differentiation of E. coli through characteristic color reactions while placed on the agar surface post-incubation. The membranes contain chromogenic substrates X-Gal, which serve as the basis for differentiation by color. E. coli cleaves X-Gal giving a blue coloration to the colony and MUG, which results in fluorescence under UV light, making it a dual detection method. The color can be visually detected on the membrane.

Application

Dual Confirmation of E. coli ID Membrane is used for rapid detection & confirmation of Escherichia coli in water and food samples, based on a chromogenic and fluorogenic method. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service