Skip to Content
Merck
All Photos(1)

Key Documents

38837

Sigma-Aldrich

Laccase, Coriolus versicolor, CLEA

≥0.3 U/mg

Synonym(s):

Laccase, Coriolus versicolor, cross-linked enzyme aggregate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

powder

Quality Level

specific activity

≥0.3 U/mg

storage temp.

2-8°C

Unit Definition

1 U corresponds to the amount of enzyme which oxidizes 1 μmol ABTS per minute at pH 4.5 and 25°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service