Skip to Content
Merck

Skip To

C3171

Dewar 35L accessories

aluminum cane with tube stops

Synonym(s):

9717089, cryo cane, cryogenic storage, dewar tube storage, tube holding cane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 ea
Check Cart for Availability
€ 199,00

About This Item

NACRES:
NB.12
UNSPSC Code:
41103000

€ 199,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pack of 50 ea

manufacturer/tradename

Worthington 9717089

diam. × L

1.2 cm × 29 cm

General description

Holds 5 tubes, each up to approx. 5 cm in height.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
R038452125BCHS120187
packaging

pack of 50 ea

packaging

pack of 1 ea

packaging

-

packaging

pk of 2

diam. × L

1.2 cm × 29 cm

diam. × L

-

diam. × L

-

diam. × L

-

manufacturer/tradename

Worthington 9717089

manufacturer/tradename

Worthington R036-9C34

manufacturer/tradename

BioControl Systems 52125, FLASH®

manufacturer/tradename

Heathrow Scientific HS120187




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library