Skip to Content
Merck
All Photos(1)

Key Documents

38155

Water

with 0.1% trifluoroacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O
Molecular Weight:
18.02
MDL number:
UNSPSC Code:
41100000
PubChem Substance ID:

Product Name

Water with 0.1% trifluoroacetic acid,

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Mary A Rodgers et al.
The Journal of experimental medicine, 211(7), 1333-1347 (2014-06-25)
Linear ubiquitination is a newly discovered posttranslational modification that is currently restricted to a small number of known protein substrates. The linear ubiquitination assembly complex (LUBAC), consisting of HOIL-1L, HOIP, and Sharpin, has been reported to activate NF-κB-mediated transcription in
M Lieveld et al.
Virchows Archiv : an international journal of pathology, 465(6), 703-713 (2014-10-12)
Giant cell tumor of bone (GCTB) displays worrisome clinical features such as local recurrence and occasionally metastatic disease which are unpredictable by morphology. Additional routinely usable biomarkers do not exist. Gene expression profiles of six clinically defined groups of GCTB
Rachele M Pojednic et al.
Calcified tissue international, 96(3), 256-263 (2014-12-07)
Vitamin D receptor (VDR) expression and action in non-human skeletal muscle have recently been reported in several studies, yet data on the activity and expression of VDR in human muscle cells are scarce. We conducted a series of studies to
Susu Duan et al.
Nature communications, 5, 5029-5029 (2014-10-10)
Oseltamivir-resistant H1N1 influenza viruses carrying the H275Y neuraminidase mutation predominated worldwide during the 2007-2009 seasons. Although several neuraminidase substitutions were found to be necessary to counteract the adverse effects of H275Y, the order and impact of evolutionary events involved remain
Lydia E H Paul et al.
Organic & biomolecular chemistry, 13(3), 946-953 (2014-11-21)
Interactions between three hexacationic arene ruthenium metallaprisms, [(p-cymene)6Ru6(tpt)2(dhnq)3](6+), [(p-cymene)6Ru6(tpt)2(dhbq)3](6+) and [(p-cymene)6Ru6(tpt)2(oxa)3](6+), and a series of human proteins including human serum albumin, transferrin, cytochrome c, myoglobin and ubiquitin have been studied using NMR spectroscopy, mass spectrometry and circular dichroism spectroscopy. All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service