Skip to Content
Merck
All Photos(1)

Key Documents

1.08178

Supelco

Trifluoroacetic acid

for protein sequence analysis

Synonym(s):

TFA

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€154.00

€154.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
50 ML
€154.00

About This Item

Linear Formula:
CF3COOH
CAS Number:
Molecular Weight:
114.02
Beilstein:
742035
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-929-3
NACRES:
NA.21

€154.00


Estimated to ship on29 May 2025


Request a Bulk Order

vapor density

3.9 (vs air)

Quality Level

vapor pressure

97.5 mmHg ( 20 °C)

Assay

≥99.7% (acidimetric)

form

liquid

impurities

≤0.02% Water (Karl Fischer)

refractive index

n20/D 1.3 (lit.)

pH

1 (10 g/L in H2O)

bp

72.4 °C (lit.)

mp

−15.4 °C (lit.)

solubility

soluble 10 g/mL

density

1.489 g/mL at 20 °C (lit.)

storage temp.

2-30°C

SMILES string

OC(C(F)(F)F)=O

InChI

1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

DTQVDTLACAAQTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trifluoroacetic acid (TFA) can be used as an ion-pairing agent in a reverse-phase HPLC system to separate peptides because it enhances peak resolution. It also has low absorption detection wavelengths and is relatively volatile and can be easily removed. For the majority of peptide separations, TFA has been used at concentrations ranging from 0.05 to 0.1%(6.5-13 mM).[1]

Analysis Note

Assay (acidimetric): ≥ 99.7 %
Identity (IR-spectrum): passes test
Density (d 20 °C/20 °C): about 1.53
Boiling point: about 72 °C
Water (according to Karl Fischer): ≤ 0.02 %

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

212.0 °F - Pensky-Martens closed cup

Flash Point(C)

> 100 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service