Skip to Content
Merck
All Photos(1)

Key Documents

37072

Supelco

Formaldehyde test kit Quantofix®

10-200 mg/L (HCHO)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

description

semi-quantitative

specific analyte(s)

formaldehyde

measuring range

10-200 mg/L (HCHO)

application(s)

environmental
food and beverages
general analytical

detection method

colorimetric

General description

Contents:
Kit including reagents and test sticks for 100 tests.

Storage:
Avoid exposing the sticks to sunlight and moisture. Store the kit below +30 °C in a dry place.

Gradation:
0 − 10 − 20 − 40 − 60 − 100 − 200 mg/L HCHO

Legal Information

Quantofix is a registered trademark of Macherey-Nagel GmbH & Co., KG

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service