Skip to Content
Merck
All Photos(2)

Key Documents

MAK140

Sigma-Aldrich

Colorimetric Aldehyde Assay Kit

sufficient for 200 colorimetric tests

Synonym(s):

Aldehyde Detection Assay Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€461.00

€461.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 KIT
€461.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

€461.00


Usually ships in 2 weeks.

Request a Bulk Order

usage

sufficient for 200 colorimetric tests

application(s)

environmental

detection method

colorimetric

relevant disease(s)

neurological disorders

storage temp.

−20°C

General description

Aldehydes are highly reactive molecules, which play a role in many physiological, pathological and commercial processes. They have been implicated as environmental pollutants, and in neurodegeneration and DNA damage. A method for rapid and accurate measurement of aldehydes is an important tool for biological and chemical research, the food industry, and environmental pollution surveillance.

Suitability

This kit is suitable for the detection of aldehydes in a variety of samples, particularly when the samples are at a lower pH. When working with samples in a high pH, product number MAK139, Colorimetric Aldehyde Assay, is more suitable.

Principle

The Colorimetric Aldehyde Assay Kit, Blue provides a simple and direct procedure for measuring aldehydes in a variety of samples. Aldehydes reacts with a sensor, resulting in a colorimetric (620 nm) product proportional to the amount of aldehyde present. The Colorimetric Aldehyde Assay Kit, Blue has a lower limit of detection of 0.3 nanomole of aldehyde in a 100 μL assay volume (3 μM).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service