Skip to Content
Merck
All Photos(2)

Key Documents

37054

Supelco

Halfenprox

PESTANAL®, analytical standard

Synonym(s):

2-(4-Bromodifluoromethoxyphenyl)-2-methylpropyl 3-phenoxybenzyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€117.00

€117.00


Estimated to ship on30 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€117.00

About This Item

Empirical Formula (Hill Notation):
C24H23BrF2O3
CAS Number:
Molecular Weight:
477.34
Beilstein:
11404200
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€117.00


Estimated to ship on30 April 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(COCC1=CC(=CC=C1)OC2=CC=CC=C2)C3=CC=C(C=C3)OC(F)(F)Br

InChI

1S/C24H23BrF2O3/c1-23(2,19-11-13-21(14-12-19)30-24(25,26)27)17-28-16-18-7-6-10-22(15-18)29-20-8-4-3-5-9-20/h3-15H,16-17H2,1-2H3

InChI key

WIFXJBMOTMKRMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Halfenprox is a pyrethroid insecticide/acaricide, found to be effective against a variety of mite species such as Tetranuchus spp. and Panonychus spp. in citrus, vegetables, fruits and ornamentals.[1]

Application

Halfenprox may be used as an analytical reference standard for the quantification of the analyte in tea[2] and food[3][4] samples using different chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimization of a multi-residue screening method for the determination of 85 pesticides in selected food matrices by stir bar sorptive extraction and thermal desorption GC-MS
Ochiai N, et al.
Journal of Separation Science, 28(9-10), 1083-1092 (2005)
The Toxicology and Biochemistry of Insecticides, Second Edition (2014)
Multiresidue method for the determination of pesticides in Oolong tea using QuEChERS by gas chromatography-triple quadrupole tandem mass spectrometry
Wu C-C
Food Chemistry, 229, 580-587 (2017)
Pesticides: Strategies for Pesticides Analysis (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service