
Halfenprox

Synonym(s):
2-(4-Bromodifluoromethoxyphenyl)-2-methylpropyl 3-phenoxybenzyl ether
Empirical Formula (Hill Notation):
C24H23BrF2O3
CAS Number:
Molecular Weight:
477.34
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2