Skip to Content
Merck
All Photos(1)

Key Documents

37019

Supelco

Atropine

analytical standard

Synonym(s):

Hyoscyamine, Tropine tropate, endo-(±)-α-(Hydroxymethyl)benzeneacetic acid 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23NO3
CAS Number:
Molecular Weight:
289.37
Beilstein:
91260
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

CN1[C@H]2CC[C@@H]1C[C@@H](C2)OC(=O)C(CO)c3ccccc3

InChI

1S/C17H23NO3/c1-18-13-7-8-14(18)10-15(9-13)21-17(20)16(11-19)12-5-3-2-4-6-12/h2-6,13-16,19H,7-11H2,1H3/t13-,14+,15+,16?

InChI key

RKUNBYITZUJHSG-SPUOUPEWSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Atropine is an antimuscarinic drug, which is classified under the tropane alkaloid family of compounds.
Atropine is an antimuscarinic drug, which is classified under the tropane alkaloid family of compounds.
For the analysis of the phytotoxins.

Application

Atropine may be used as a reference standard for the determination of atropine in:
  • Blood samples by liquid chromatography combined with electrospray ionization-triple quadrupole mass spectrometry (LC-ESI-MS/MS).
  • Hair samples by ultra-high performance liquid chromatography-high resolution mass spectrometry (UHPLC-HRMS) consisting of single-stage Orbitrap MS system with ESI interface.
  • Oganic buckwheat (Fagopyron esculentum L.) products by UHPLC-MS/MS.
  • Plant materials by liquid-liquid extraction and EXtrelut® column clean-up followed by analysis using gas chromatography (GC) coupled with flame ionization detector (FID) and MS.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Atropine may be used as a reference standard for the determination of atropine in:
  • Blood samples by liquid chromatography combined with electrospray ionization-triple quadrupole mass spectrometry (LC-ESI-MS/MS).
  • Hair samples by ultra-high performance liquid chromatography-high resolution mass spectrometry (UHPLC-HRMS) consisting of single-stage Orbitrap MS system with ESI interface.
  • Oganic buckwheat (Fagopyron esculentum L.) products by UHPLC-MS/MS.
  • Plant materials by liquid-liquid extraction and EXtrelut® column clean-up followed by analysis using gas chromatography (GC) coupled with flame ionization detector (FID) and MS.

Biochem/physiol Actions

Competitive nonselective antagonist at central and peripheral muscarinic acetylcholine receptors.

Reconstitution

Dried down, ~100μg/mL after reconstitution with 1 mL of water

Analysis Note

purity : ≥96.0% (HPLC)

Legal Information

Extrelut is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Are tropane alkaloids present in organic foods? Detection of scopolamine and atropine in organic buckwheat (Fagopyron esculentum L.) products by UHPLC-MS/MS.
<BIG>Cirlini M, et al.</BIG>
Food Chemistry, 239, 141-147 (2018)
High-throughput screening for drugs of abuse and pharmaceutical drugs in hair by liquid-chromatography-high resolution mass spectrometry (LC-HRMS).
<BIG>Odoardi S, et al.</BIG>
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 133, 302-310 (2017)
Enantiomeric determination and evaluation of the racemization process of atropine in Solanaceae seeds and contaminated samples by high performance liquid chromatography-tandem mass spectrometry.
<BIG>Marin-Saez J, et al.</BIG>
Journal of Chromatography A, 1474, 79-84 (2016)
Isolation of atropine and scopolamine from plant material using liquid-liquid extraction and EXtrelut columns
<BIG>Sramska P, et al.</BIG>
Journal of Chromatography. B, Biomedical Sciences and Applications, 1043, 202-208 (2017)
Method validation for simultaneous determination of atropine, pralidoxime and 12 organophosphorus compounds in blood samples by means of high-performance liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS).
<BIG>Marques GLM, et al</BIG>.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1097, 44-53 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service