Skip to Content
Merck
All Photos(2)

Key Documents

P49805

Sigma-Aldrich

Piperonylic acid

99%

Synonym(s):

1,3-Benzodioxole-5-carboxylic acid, 3,4-(Methylenedioxy)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6O4
CAS Number:
Molecular Weight:
166.13
Beilstein:
150206
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

powder

mp

229-231 °C (lit.)

SMILES string

OC(=O)c1ccc2OCOc2c1

InChI

1S/C8H6O4/c9-8(10)5-1-2-6-7(3-5)12-4-11-6/h1-3H,4H2,(H,9,10)

InChI key

VDVJGIYXDVPQLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Ranga Rao et al.
Bioorganic & medicinal chemistry, 17(14), 5170-5175 (2009-06-12)
A bioassay-guided fractionation and chemical examination of antihyperglycemic root extract of Derris indica resulted in isolation and characterization of two new furanoflavanoids (1, 2) along with thirteen known compounds (3-15). Their structures were determined on the basis of extensive spectroscopic
Guillaume A Schoch et al.
Plant physiology, 130(2), 1022-1031 (2002-10-12)
The cinnamate (CA) 4-hydroxylase (C4H) is a cytochrome P450 that catalyzes the second step of the main phenylpropanoid pathway, leading to the synthesis of lignin, pigments, and many defense molecules. Salicylic acid (SA) is an essential trigger of plant disease
Yue Zhu et al.
Journal of the American Chemical Society, 128(13), 4267-4276 (2006-03-30)
Two-photon excitation (2PE) of "caged" biomolecules represents a powerful method to investigate the temporal and spatial relevance of physiological function in real time and on living tissue, because the excitation volume can be restricted to 1 fL. Additionally, low-energy IR
Debabrata Sircar et al.
Journal of plant physiology, 166(13), 1370-1380 (2009-04-04)
Biosynthesis of hydroxybenzoates even at enzymatic level is poorly understood. In this report, effect of feeding of putative biosynthetic precursors and pathway-specific enzyme inhibitors of early phenylpropanoid pathway on p-hydroxybenzoic acid accumulation in chitosan-elicited hairy roots of Daucus carota was
Eyal Shimoni et al.
Journal of biotechnology, 105(1-2), 61-70 (2003-09-27)
Propenylbenzenes are often used as starting materials in the chemical synthesis of aroma compounds and fine chemicals. In the present study, we demonstrate the ability of an Arthrobacter sp. to transform various structures of propenylbenzenes derived from essential oils to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service